Gene Transcripts

Gene Symbol
RLN2
Entrez Gene ID
6019
Gene Name
relaxin 2
Gene Aliases
H2, H2-RLX, RLXH2, bA12D24.1.1, bA12D24.1.2
Chromosome Location
Chr. 9: 5299864 - 5306134 on Build GRCh38
UniGene ID
Hs.127032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005059.3 NP_005050.2 1 119
NM_134441.2 NP_604390.1 1 119
XM_017015011.1 XP_016870500.1 2 488
GenBank mRNA A06925.1 - 1 88
A17315.1 - 1 88
BC126415.1 - 1 119 8992131
BC126419.1 - 1 119 8992135
X00948.1 - 1 88

View Full Product Details