Gene Transcripts

Gene Symbol
Nos2
Entrez Gene ID
24599
Gene Name
nitric oxide synthase 2, inducible
Gene Aliases
Nos2a, iNos
Chromosome Location
Chr. 10: 66188290 - 66221621 on Build Rnor_6.0
UniGene ID
Rn.10400
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012611.3 NP_036743.3 1648
GenBank mRNA AF006619.1 - 1 185
AF006620.1 - 1 185
AY211532.1 - 1459
D12520.1 - 1648
D14051.1 - 1478
D44591.1 - 1479
D83661.1 - 1458
L12562.1 - 1560
S71597.1 - 1 24
U03699.1 - 1461
U26686.1 - 1458
X76881.1 - 1643

Gene Symbol
LOC497963
Entrez Gene ID
497963
Gene Name
similar to Nitric oxide synthase, inducible (NOS type II) (Inducible NO synthase) (Inducible NOS) (iNOS)
Gene Aliases
Chromosome Location
Chr. 10: 66282166 - 66313364 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246949.2 XP_006247011.1 1513

View Full Product Details