Gene Transcripts
- Gene Symbol
- Gfer
- Entrez Gene ID
- 27100
- Gene Name
- growth factor, augmenter of liver regeneration
- Gene Aliases
- ALR
- Chromosome Location
- Chr. 10: 14059347 - 14061703 on Build Rnor_6.0
- UniGene ID
- Rn.11039
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013222.2 |
NP_037354.2
|
3 |
589 |
|
GenBank mRNA |
D30735.1 |
- |
3 |
589 |
|
View Full Product Details