Gene Transcripts

Gene Symbol
Pde2a
Entrez Gene ID
81743
Gene Name
phosphodiesterase 2A, cGMP-stimulated
Gene Aliases
Pde2, Pde2a2
Chromosome Location
Chr. 1: 166534643 - 166626158 on Build Rnor_6.0
UniGene ID
Rn.10044
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143847.1 NP_001137319.1 20, 21 1909
NM_001270604.1 NP_001257533.1 20, 21 1859
NM_031079.1 NP_112341.1 19, 20 1734
GenBank mRNA U21101.1 - 19, 20 1734

View Full Product Details