Gene Transcripts

Gene Symbol
Gps1
Entrez Gene ID
117039
Gene Name
G protein pathway suppressor 1
Gene Aliases
MFH
Chromosome Location
Chr. 10: 109966875 - 109971912 on Build Rnor_6.0
UniGene ID
Rn.16873
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270939.1 NP_001257868.1 13, 14 1407
NM_053969.3 NP_446421.3 13, 14 1581
XM_008768462.1 XP_008766684.1 13, 14 1573
XM_008768463.1 XP_008766685.1 13, 14 1433
GenBank mRNA BC061746.1 - 13, 14 1530
X87885.1 - 12, 13 1362

View Full Product Details