Gene Transcripts

Gene Symbol
Pkn2
Entrez Gene ID
207122
Gene Name
protein kinase N2
Gene Aliases
Pak-2, Prkcl2
Chromosome Location
Chr. 2: 248735699 - 248840735 on Build Rnor_6.0
UniGene ID
Rn.30325
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105755.2 NP_001099225.2 7, 8 1518
XM_008761494.1 XP_008759716.1 7, 8 1373
GenBank mRNA U75358.1 - 5, 6 757

View Full Product Details