Gene Transcripts

Gene Symbol
Qtrtd1
Entrez Gene ID
288364
Gene Name
queuine tRNA-ribosyltransferase domain containing 1
Gene Aliases
Chromosome Location
Chr. 11: 61748842 - 61779631 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006248320.2 XP_006248382.1 9 1186
XM_008768759.1 XP_008766981.1 9 1225
XM_008768760.1 XP_008766982.1 8 1012
XM_008768761.1 XP_008766983.1 8 973
XM_008768762.1 XP_008766984.1 7 843

View Full Product Details