Gene Transcripts

Gene Symbol
Sympk
Entrez Gene ID
292683
Gene Name
symplekin
Gene Aliases
Hfn3g
Chromosome Location
Chr. 1: 79930906 - 79959461 on Build Rnor_6.0
UniGene ID
Rn.44851
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100830.1 NP_001094300.1 18 2476
XM_008758908.1 XP_008757130.1 18 2726

View Full Product Details