Gene Transcripts

Gene Symbol
Slc39a2
Entrez Gene ID
305846
Gene Name
solute carrier family 39 (zinc transporter), member 2
Gene Aliases
Chromosome Location
Chr. 15: 28295057 - 28297688 on Build Rnor_6.0
UniGene ID
Rn.51972
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107260.1 NP_001100730.1 1 209
XM_008770521.1 XP_008768743.1 1 728

View Full Product Details