Gene Transcripts

Gene Symbol
Txndc15
Entrez Gene ID
307180
Gene Name
thioredoxin domain containing 15
Gene Aliases
RGD1304696
Chromosome Location
Chr. 17: 9429171 - 9441628 on Build Rnor_6.0
UniGene ID
Rn.233757
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024998.1 NP_001020169.1 3 625
XM_008771497.1 XP_008769719.1 3 733
GenBank mRNA BC091340.1 - 3 625

View Full Product Details