Gene Transcripts

Gene Symbol
IFITM1
Entrez Gene ID
8519
Gene Name
interferon induced transmembrane protein 1
Gene Aliases
9-27, CD225, DSPA2a, IFI17, LEU13
Chromosome Location
Chr. 11: 313991 - 315272 on Build GRCh38
UniGene ID
Hs.458414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003641.3 NP_003632.3 1 250
GenBank mRNA AK290480.1 - 1 225
BC000897.1 - 1 200 3464598
BT007173.1 - 1 70
J04164.1 - 1 386

View Full Product Details