Gene Transcripts

Gene Symbol
Krt78
Entrez Gene ID
315324
Gene Name
keratin 78, type II
Gene Aliases
Kb40
Chromosome Location
Chr. 7: 143545245 - 143552723 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765864.1 XP_008764086.1 4 811

View Full Product Details