Gene Transcripts

Gene Symbol
Zdhhc18
Entrez Gene ID
362613
Gene Name
zinc finger, DHHC-type containing 18
Gene Aliases
Chromosome Location
Chr. 5: 151829795 - 151856256 on Build Rnor_6.0
UniGene ID
Rn.203345
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039339.1 NP_001034428.1 4 728
GenBank mRNA AY886534.1 - 4 728

View Full Product Details