Gene Transcripts

Gene Symbol
Ctdnep1
Entrez Gene ID
67181
Gene Name
CTD nuclear envelope phosphatase 1
Gene Aliases
2610507E10Rik, Dullard
Chromosome Location
Chr. 11: 69981168 - 69990601 on Build GRCm38
UniGene ID
Mm.41678
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026017.2 NP_080293.1 3 540
GenBank mRNA AK012063.1 - 3 522
AK076150.1 - 3 554
AK164602.1 - 3 482
BC018265.1 - 3 540 4486464

View Full Product Details