Gene Transcripts

Gene Symbol
Grrp1
Entrez Gene ID
72690
Gene Name
glycine/arginine rich protein 1
Gene Aliases
2810043G13Rik, Fam110d
Chromosome Location
Chr. 4: 134251102 - 134266197 on Build GRCm38
UniGene ID
Mm.100168
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099296.2 NP_001092766.1 2 659
GenBank mRNA AK012898.1 - 2 650
BC048704.1 - 2 655 6330338

View Full Product Details