Gene Transcripts

Gene Symbol
RAB30
Entrez Gene ID
27314
Gene Name
RAB30, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 11: 82973133 - 83071923 on Build GRCh38
UniGene ID
Hs.40758
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286059.1 NP_001272988.1 4 619
NM_001286060.1 NP_001272989.1 4 454
NM_001286061.1 NP_001272990.1 4 352
NM_014488.4 NP_055303.2 5 555
GenBank mRNA AF498956.1 - 3 213
AK291376.1 - 4 614
AK310082.1 - 2 226
AK312323.1 - 4 360
BC014213.2 - 5 474 4763140
BP336371.1 - 4 352
BX641138.1 - 4 403
CR542106.1 - 3 213
DB151524.1 - 4 455
U57092.1 - 3 231

View Full Product Details