Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001383.3 | NP_001374.3 | 8 | 805 | |
GenBank mRNA | AF321876.1 | - | 8 | 805 | |
AK090530.1 | - | 2 | 878 | ||
AK297947.1 | - | 7 | 823 | ||
AK314015.1 | - | 8 | 791 | ||
BC003099.1 | - | 8 | 767 | ||
BC096088.1 | - | 7 | 661 | 40000799 | |
BT019878.1 | - | 6 | 547 | ||
S81752.1 | - | 8 | 840 | ||
U34880.1 | - | 8 | 840 |