Gene Transcripts

Gene Symbol
DPH1
Entrez Gene ID
1801
Gene Name
diphthamide biosynthesis 1
Gene Aliases
DEDSSH, DPH2L, DPH2L1, OVCA1
Chromosome Location
Chr. 17: 2030137 - 2043431 on Build GRCh38
UniGene ID
Hs.513856
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001383.3 NP_001374.3 9 1021
GenBank mRNA AK297947.1 - 8 1039
BP417755.1 - 1 70

View Full Product Details