Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243325.1 | NP_001230254.1 | 1 | 202 | |
NM_001243327.1 | NP_001230256.1 | 1 | 202 | ||
NM_001243328.1 | NP_001230257.1 | 2 | 416 | ||
NM_139165.2 | NP_631904.1 | 1 | 202 | ||
XM_011535475.2 | XP_011533777.1 | 3 | 722 | ||
XM_011535476.2 | XP_011533778.1 | 3 | 736 | ||
XM_011535478.2 | XP_011533780.1 | 3 | 719 | ||
XM_011535479.2 | XP_011533781.1 | 3 | 721 | ||
XM_017010285.1 | XP_016865774.1 | 3 | 721 | ||
XM_017010286.1 | XP_016865775.1 | 3 | 721 | ||
XM_017010287.1 | XP_016865776.1 | 3 | 721 | ||
XM_017010288.1 | XP_016865777.1 | 1 | 74 | ||
GenBank mRNA | AF359243.1 | - | 1 | 90 | |
AY054974.1 | - | 1 | 69 | ||
AY069961.1 | - | 1 | 129 | ||
AY176317.1 | - | 1 | 69 | ||
AY252119.1 | - | 1 | 69 | ||
AY359075.1 | - | 2 | 416 | ||
BC101976.1 | - | 1 | 100 | ||
BC103694.1 | - | 1 | 100 | 40003751 | |
EF489426.1 | - | 1 | 69 |