Gene Transcripts

Gene Symbol
EPHB3
Entrez Gene ID
2049
Gene Name
EPH receptor B3
Gene Aliases
EK2, ETK2, HEK2, TYRO6
Chromosome Location
Chr. 3: 184561799 - 184582408 on Build GRCh38
UniGene ID
Hs.2913
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004443.3 NP_004434.2 12 2809
GenBank mRNA BC052968.1 - 12 2792
X75208.1 - 12 2399

View Full Product Details