Gene Transcripts

Gene Symbol
BRICD5
Entrez Gene ID
283870
Gene Name
BRICHOS domain containing 5
Gene Aliases
C16orf79
Chromosome Location
Chr. 16: 2209253 - 2213037 on Build GRCh38
UniGene ID
Hs.647399
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182563.3 NP_872369.2 6 850
XM_005255263.4 XP_005255320.1 5 2915
GenBank mRNA AY597811.1 - 5 711
BC039154.2 - 6 792 4213860

View Full Product Details