Gene Transcripts

Gene Symbol
OOEP
Entrez Gene ID
441161
Gene Name
oocyte expressed protein
Gene Aliases
C6orf156, FLOPED, HOEP19, KHDC2
Chromosome Location
Chr. 6: 73368553 - 73369888 on Build GRCh38
UniGene ID
Hs.671212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080507.2 NP_001073976.1 3 607
XM_017010890.1 XP_016866379.1 3 586

View Full Product Details