Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000402.4 | NP_000393.4 | 9 | 1268 | |
NM_001042351.2 | NP_001035810.1 | 9 | 1157 | ||
GenBank mRNA | AK292304.1 | - | 9 | 1094 | |
AK302341.1 | - | 9 | 1095 | ||
BC000337.2 | - | 9 | 1083 | 2822640 | |
M21248.1 | - | 8 | 1044 | ||
X03674.1 | - | 9 | 1500 |