Gene Transcripts

Gene Symbol
Bub1b
Entrez Gene ID
12236
Gene Name
budding uninhibited by benzimidazoles 1 homolog, beta (S. cerevisiae)
Gene Aliases
AU045529, BUBR1
Chromosome Location
Chr. 2: 118598203 - 118641592 on Build GRCm38
UniGene ID
Mm.29133
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009773.3 NP_033903.2 9 1471
GenBank mRNA AF107296.1 - 9 1426
AK133909.1 - 9 1412
BC031577.1 - 9 1422 4953223

View Full Product Details