Gene Transcripts

Gene Symbol
Dsg3
Entrez Gene ID
13512
Gene Name
desmoglein 3
Gene Aliases
bal, balding
Chromosome Location
Chr. 18: 20510304 - 20541310 on Build GRCm38
UniGene ID
Mm.106811
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030596.3 NP_085099.2 16 3008
GenBank mRNA AK037142.1 - 16 3039
AK148657.1 - 16 3009
BC132058.1 - 16 3037 40130335
BC132639.1 - 16 3037 40130916
U86016.1 - 16 2937

View Full Product Details