Gene Transcripts

Gene Symbol
Dsg3
Entrez Gene ID
13512
Gene Name
desmoglein 3
Gene Aliases
bal, balding
Chromosome Location
Chr. 18: 20510304 - 20541310 on Build GRCm38
UniGene ID
Mm.106811
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030596.3 NP_085099.2 16 2582
GenBank mRNA AK029018.1 - 2604
AK037142.1 - 16 2613
AK148657.1 - 16 2583
BC132058.1 - 16 2611 40130335
BC132639.1 - 16 2611 40130916
U86016.1 - 16 2511

View Full Product Details