Gene Transcripts

Gene Symbol
Ephb3
Entrez Gene ID
13845
Gene Name
Eph receptor B3
Gene Aliases
AW456895, Cek10, Etk2, HEK2, MDK5, Sek4, Tyro6
Chromosome Location
Chr. 16: 21204795 - 21223309 on Build GRCm38
UniGene ID
Mm.6972
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010143.1 NP_034273.1 3 696
GenBank mRNA AK144414.1 - 3 712
AK158043.1 - 3 1757
BC014822.1 - 3 697 3673003
BC053085.1 - 3 696 5708827
U11493.1 - 1 156
Z49086.1 - 3 554

View Full Product Details