Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001285457.1 | NP_001272386.1 | 2 | 540 | |
NM_010279.3 | NP_034409.1 | 3 | 741 | ||
GenBank mRNA | AB000800.1 | - | 2 | 295 | |
AF015172.1 | - | 2 | 523 | ||
AK076171.1 | - | 3 | 661 | ||
BC011532.1 | - | 2 | 486 | ||
BC040251.1 | - | 2 | 324 | ||
BC044783.2 | - | 3 | 620 | ||
BC054378.1 | - | 2 | 412 | 6390018 | |
KM079499.1 | - | 2 | 400 |