Gene Transcripts

Gene Symbol
Klra3
Entrez Gene ID
16634
Gene Name
killer cell lectin-like receptor, subfamily A, member 3
Gene Aliases
5E6, Ly49c, NK-2.1, Nk-2, Nk2, Nk2.1
Chromosome Location
Chr. 6: 130323289 - 130337626 on Build GRCm38
UniGene ID
Mm.425338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289604.1 NP_001276533.1 4 560
NM_001289605.1 NP_001276534.1 4 594
NM_010648.3 NP_034778.2 4 560
GenBank mRNA AK159595.1 - 4 509
BC099867.1 - 392 40040425
BC099878.1 - 392 40040424
BC099879.1 - 392 40040423
BC099880.1 - 392 40040422
BC101952.1 - 4 436 40044699
BC103543.1 - 4 436 40044703
BC103544.1 - 4 470 40044701
BC103545.1 - 4 436 40044700
BC132524.1 - 4 441 40130801
BC132526.1 - 4 441 40130803
BC145192.1 - 4 441 9053743
BC145193.1 - 4 475 9053744
U09739.1 - 4 560
U10305.1 - 4 476
U49868.1 - 4 541
U56404.1 - 4 435
U56405.1 - 4 435

View Full Product Details