Gene Transcripts

Gene Symbol
Olfr17
Entrez Gene ID
18314
Gene Name
olfactory receptor 17
Gene Aliases
MOR263-5, P2
Chromosome Location
Chr. 7: 107097467 - 107098414 on Build GRCm38
UniGene ID
Mm.103814
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020598.2 NP_065623.2 1 396
GenBank mRNA BC139721.1 - 1 396

View Full Product Details