Gene Transcripts

Gene Symbol
Rab30
Entrez Gene ID
75985
Gene Name
RAB30, member RAS oncogene family
Gene Aliases
5033421K01Rik, AI323892, Rsb30
Chromosome Location
Chr. 7: 92741714 - 92844535 on Build GRCm38
UniGene ID
Mm.323004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029494.2 NP_083770.2 3, 4 307
GenBank mRNA AB232626.1 - 2, 3 170
AF399756.1 - 2, 3 170
AK017185.1 - 2, 3 257
AK142206.1 - 3, 4 314
BC017550.1 - 3, 4 307 4503909

View Full Product Details