Gene Transcripts

Gene Symbol
Tnfrsf23
Entrez Gene ID
79201
Gene Name
tumor necrosis factor receptor superfamily, member 23
Gene Aliases
SOB, TNFRSFH23, Tnfrh1, Tnfrsf1al1, mDcTRAILR1, mSOB
Chromosome Location
Chr. 7: 143665807 - 143686423 on Build GRCm38
UniGene ID
Mm.290780
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ278264.1 - 2 82
AK154786.1 - 2 108
AK170037.1 - 2 111
AY046550.1 - 2 91
AY165625.1 - 2 97
BC106828.1 - 2 93 40044453
BC106829.1 - 2 97 40044455
BG077775.2 - 2 83
BY036522.1 - 2 107
BY201915.1 - 2 113

View Full Product Details