Gene Transcripts

Gene Symbol
FGFR3
Entrez Gene ID
2261
Gene Name
fibroblast growth factor receptor 3
Gene Aliases
ACH, CD333, CEK2, HSFGFR3EX, JTK4
Chromosome Location
Chr. 4: 1793299 - 1808872 on Build GRCh38
UniGene ID
Hs.1420
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000142.4 NP_000133.1 18 3264
NM_001163213.1 NP_001156685.1 18 3270
NM_022965.3 NP_075254.1 16 2928
XM_006713868.1 XP_006713931.1 18 3292
XM_006713869.1 XP_006713932.1 18 3289
XM_006713870.1 XP_006713933.1 18 3289
XM_006713871.1 XP_006713934.1 18 3286
XM_006713872.1 XP_006713935.1 18 3280
XM_006713873.1 XP_006713936.1 18 3280
XM_011513420.1 XP_011511722.1 18 3286
XM_011513422.1 XP_011511724.1 18 3283
GenBank mRNA AB209441.1 - 17 3199
M64347.1 - 16 2778

View Full Product Details