Gene Transcripts
- Gene Symbol
- Galnt13
- Entrez Gene ID
- 271786
- Gene Name
- UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13
- Gene Aliases
- A230002A12, A230020F20, BB182356
- Chromosome Location
- Chr. 2: 54436387 - 55118309 on Build GRCm38
- UniGene ID
- Mm.330227
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038387.1 |
- |
11 |
1913 |
|
View Full Product Details