Gene Transcripts

Gene Symbol
Pgm5
Entrez Gene ID
226041
Gene Name
phosphoglucomutase 5
Gene Aliases
4833423B07, 9530034F03Rik, D830025G17, aciculin
Chromosome Location
Chr. 19: 24678261 - 24861842 on Build GRCm38
UniGene ID
Mm.105222
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175013.2 NP_778178.3 5 865
GenBank mRNA AK035507.1 - 5 865
AK076484.1 - 4 564
BC115937.1 - 5 675 40089650

View Full Product Details