Gene Transcripts
- Gene Symbol
- Sclt1
- Entrez Gene ID
- 67161
- Gene Name
- sodium channel and clathrin linker 1
- Gene Aliases
- 2610207F23Rik, 4931421F20Rik
- Chromosome Location
- Chr. 3: 41626705 - 41742530 on Build GRCm38
- UniGene ID
- Mm.331001
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC043136.1 |
- |
4, 5 |
680 |
|
BC060271.1 |
- |
4, 5 |
680 |
6812105 |
View Full Product Details