Gene Transcripts

Gene Symbol
GTDC1
Entrez Gene ID
79712
Gene Name
glycosyltransferase like domain containing 1
Gene Aliases
Hmat-Xa, mat-Xa
Chromosome Location
Chr. 2: 143945787 - 144333092 on Build GRCh38
UniGene ID
Hs.44780
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006636.3 NP_001006637.1 7 974
NM_001164629.3 NP_001158101.1 7 959
NM_001284233.1 NP_001271162.1 5 783
NM_001284234.1 NP_001271163.1 5 674
NM_001284235.1 NP_001271164.1 7 941
NM_001284238.1 NP_001271167.1 9 1173
NM_024659.4 NP_078935.2 7 941
XM_005263774.2 XP_005263831.1 8 1058
XM_005263782.2 XP_005263839.1 8 1074
XM_005263784.2 XP_005263841.1 6 933
XM_011511843.2 XP_011510145.1 8 1107
XM_011511855.2 XP_011510157.1 6 1051
XM_011511856.2 XP_011510158.1 6 833
XM_017004917.1 XP_016860406.1 12 1729
XM_017004918.1 XP_016860407.1 10 1321
XM_017004919.1 XP_016860408.1 8 1207
XM_017004920.1 XP_016860409.1 10 4363
XM_017004921.1 XP_016860410.1 9 1156
XM_017004922.1 XP_016860411.1 9 1333
XM_017004923.1 XP_016860412.1 8 1121
XM_017004924.1 XP_016860413.1 10 1317
XM_017004925.1 XP_016860414.1 7 979
XM_017004926.1 XP_016860415.1 5 910
XM_017004927.1 XP_016860416.1 7 941
XM_017004928.1 XP_016860417.1 9 1174
XM_017004929.1 XP_016860418.1 7 911
XM_017004930.1 XP_016860419.1 7 1562
XM_017004931.1 XP_016860420.1 7 975
XM_017004932.1 XP_016860421.1 7 945
XM_017004933.1 XP_016860422.1 7 927
XM_017004934.1 XP_016860423.1 8 4134
XM_017004935.1 XP_016860424.1 9 2004
XM_017004936.1 XP_016860425.1 8 1181
XM_017004937.1 XP_016860426.1 6 1062
XM_017004938.1 XP_016860427.1 6 1060
XM_017004939.1 XP_016860428.1 8 1110
XM_017004940.1 XP_016860429.1 5 911
XM_017004941.1 XP_016860430.1 5 910
XM_017004942.1 XP_016860431.1 7 957
XM_017004943.1 XP_016860432.1 7 1623
XM_017004944.1 XP_016860433.1 7 956
XM_017004945.1 XP_016860434.1 9 1174
XM_017004946.1 XP_016860435.1 7 911
XM_017004947.1 XP_016860436.1 7 1594
XM_017004948.1 XP_016860437.1 8 4134
GenBank mRNA AB201269.1 - 4 695
AB597184.1 - 4 695
AK021815.1 - 1 150
AK091684.1 - 5 787
AK291357.1 - 7 889
AY281366.1 - 7 974
BC017741.2 - 8 981 4390876
BC061699.1 - 7 881 4608518
BX431825.2 - 1 97
BX647436.1 - 5 656

View Full Product Details