Gene Transcripts
- Gene Symbol
- MUC2
- Entrez Gene ID
- 4583
- Gene Name
- mucin 2, oligomeric mucus/gel-forming
- Gene Aliases
- MLP, MUC-2, SMUC
- Chromosome Location
- Chr. 11: 1074875 - 1110508 on Build GRCh38
- UniGene ID
- Hs.315
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002457.3 |
NP_002448.3
|
|
14772 |
|
View Full Product Details