Gene Transcripts

Gene Symbol
MUC2
Entrez Gene ID
4583
Gene Name
mucin 2, oligomeric mucus/gel-forming
Gene Aliases
MLP, MUC-2, SMUC
Chromosome Location
Chr. 11: 1074875 - 1110508 on Build GRCh38
UniGene ID
Hs.315
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002457.3 NP_002448.3 13005
GenBank mRNA L21998.1 - 13005
M22405.1 - 2 758
M94132.1 - 2 393

View Full Product Details