Gene Transcripts
- Gene Symbol
- MON2
- Entrez Gene ID
- 23041
- Gene Name
- MON2 homolog, regulator of endosome-to-Golgi trafficking
- Gene Aliases
- Chromosome Location
- Chr. 12: 62466817 - 62598956 on Build GRCh38
- UniGene ID
- Hs.389378
- Species
- Homo sapiens
- Gene Symbol
- ATM
- Entrez Gene ID
- 472
- Gene Name
- ATM serine/threonine kinase
- Gene Aliases
- AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
- Chromosome Location
- Chr. 11: 108222484 - 108369102 on Build GRCh38
- UniGene ID
- Hs.367437
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC007023.2 |
- |
4 |
1228 |
|
- Gene Symbol
- BHLHE40-AS1
- Entrez Gene ID
- 100507582
- Gene Name
- BHLHE40 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 4896809 - 4979961 on Build GRCh38
- UniGene ID
- Hs.434505
- Species
- Homo sapiens
- Gene Symbol
- C15orf57
- Entrez Gene ID
- 90416
- Gene Name
- chromosome 15 open reading frame 57
- Gene Aliases
- CCDC32
- Chromosome Location
- Chr. 15: 40531341 - 40565057 on Build GRCh38
- UniGene ID
- Hs.654661
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL832032.2 |
- |
5 |
2523 |
|
- Gene Symbol
- CHAF1B
- Entrez Gene ID
- 8208
- Gene Name
- chromatin assembly factor 1 subunit B
- Gene Aliases
- CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7
- Chromosome Location
- Chr. 21: 36380145 - 36416827 on Build GRCh38
- UniGene ID
- Hs.75238
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092781.1 |
- |
1 |
1259 |
|
- Gene Symbol
- CTCF
- Entrez Gene ID
- 10664
- Gene Name
- CCCTC-binding factor
- Gene Aliases
- MRD21
- Chromosome Location
- Chr. 16: 67562407 - 67639185 on Build GRCh38
- UniGene ID
- Hs.368367
- Species
- Homo sapiens
- Gene Symbol
- CYB561A3
- Entrez Gene ID
- 220002
- Gene Name
- cytochrome b561 family member A3
- Gene Aliases
- CYBASC3, LCYTB
- Chromosome Location
- Chr. 11: 61348748 - 61362299 on Build GRCh38
- UniGene ID
- Hs.22546
- Species
- Homo sapiens
- Gene Symbol
- CYB5R2
- Entrez Gene ID
- 51700
- Gene Name
- cytochrome b5 reductase 2
- Gene Aliases
- B5R.2
- Chromosome Location
- Chr. 11: 7665095 - 7678568 on Build GRCh38
- UniGene ID
- Hs.414362
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209000.1 |
- |
2 |
2914 |
|
- Gene Symbol
- FASTKD5
- Entrez Gene ID
- 60493
- Gene Name
- FAST kinase domains 5
- Gene Aliases
- dJ1187M17.5
- Chromosome Location
- Chr. 20: 3146519 - 3159886 on Build GRCh38
- UniGene ID
- Hs.741168
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK027215.1 |
- |
1 |
1559 |
|
- Gene Symbol
- HAUS3
- Entrez Gene ID
- 79441
- Gene Name
- HAUS augmin like complex subunit 3
- Gene Aliases
- C4orf15, IT1, dgt3
- Chromosome Location
- Chr. 4: 2231836 - 2242164 on Build GRCh38
- UniGene ID
- Hs.665869
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR749640.1 |
- |
5 |
2405 |
|
- Gene Symbol
- HERC4
- Entrez Gene ID
- 26091
- Gene Name
- HECT and RLD domain containing E3 ubiquitin protein ligase 4
- Gene Aliases
- Chromosome Location
- Chr. 10: 67921899 - 68075348 on Build GRCh38
- UniGene ID
- Hs.51891
- Species
- Homo sapiens
- Gene Symbol
- KDELC2
- Entrez Gene ID
- 143888
- Gene Name
- KDEL motif containing 2
- Gene Aliases
- Chromosome Location
- Chr. 11: 108472106 - 108498432 on Build GRCh38
- UniGene ID
- Hs.83286
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125519.1 |
- |
6 |
1169 |
|
- Gene Symbol
- KIAA1671
- Entrez Gene ID
- 85379
- Gene Name
- KIAA1671
- Gene Aliases
- Chromosome Location
- Chr. 22: 24952688 - 25197448 on Build GRCh38
- UniGene ID
- Hs.419171
- Species
- Homo sapiens
- Gene Symbol
- KMT2E
- Entrez Gene ID
- 55904
- Gene Name
- lysine methyltransferase 2E
- Gene Aliases
- HDCMC04P, MLL5, NKp44L
- Chromosome Location
- Chr. 7: 105014190 - 105114085 on Build GRCh38
- UniGene ID
- Hs.592262
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY211525.1 |
- |
4 |
286 |
|
- Gene Symbol
- LINC01299
- Entrez Gene ID
- 286186
- Gene Name
- long intergenic non-protein coding RNA 1299
- Gene Aliases
- Chromosome Location
- Chr. 8: 65527008 - 65562666 on Build GRCh38
- UniGene ID
- Hs.449427
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033893.1 |
|
3 |
226 |
|
GenBank mRNA |
AK094960.1 |
- |
3 |
212 |
|
DA397003.1 |
- |
3 |
226 |
|
- Gene Symbol
- LOC100287042
- Entrez Gene ID
- 100287042
- Gene Name
- uncharacterized LOC100287042
- Gene Aliases
- Chromosome Location
- Chr. 17: 75271299 - 75273895 on Build GRCh38
- UniGene ID
- Hs.514470
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036520.1 |
|
1 |
2515 |
|
- Gene Symbol
- LOC105372392
- Entrez Gene ID
- 105372392
- Gene Name
- uncharacterized LOC105372392
- Gene Aliases
- Chromosome Location
- Chr. 19: 37328188 - 37330124 on Build GRCh38
- UniGene ID
- Hs.640243
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123379.1 |
- |
1 |
134 |
|
- Gene Symbol
- MACROD2
- Entrez Gene ID
- 140733
- Gene Name
- MACRO domain containing 2
- Gene Aliases
- C20orf133
- Chromosome Location
- Chr. 20: 13995476 - 16053197 on Build GRCh38
- UniGene ID
- Hs.661576
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC036403.2 |
- |
3 |
1318 |
|
- Gene Symbol
- MARVELD2
- Entrez Gene ID
- 153562
- Gene Name
- MARVEL domain containing 2
- Gene Aliases
- DFNB49, MARVD2, MRVLDC2, Tric
- Chromosome Location
- Chr. 5: 69415112 - 69444022 on Build GRCh38
- UniGene ID
- Hs.657687
- Species
- Homo sapiens
- Gene Symbol
- MFSD14A
- Entrez Gene ID
- 64645
- Gene Name
- major facilitator superfamily domain containing 14A
- Gene Aliases
- HIAT1
- Chromosome Location
- Chr. 1: 100038078 - 100083377 on Build GRCh38
- UniGene ID
- Hs.124156
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127671.1 |
- |
1 |
200 |
|
- Gene Symbol
- NAA40
- Entrez Gene ID
- 79829
- Gene Name
- N(alpha)-acetyltransferase 40, NatD catalytic subunit
- Gene Aliases
- NAT11, PATT1
- Chromosome Location
- Chr. 11: 63938970 - 63957322 on Build GRCh38
- UniGene ID
- Hs.523753
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX647309.1 |
- |
2 |
563 |
|
- Gene Symbol
- P2RX5-TAX1BP3
- Entrez Gene ID
- 100533970
- Gene Name
- P2RX5-TAX1BP3 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 17: 3662893 - 3696404 on Build GRCh38
- UniGene ID
- Hs.731607
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037928.1 |
|
12 |
4020 |
|
GenBank mRNA |
AB209623.1 |
- |
12 |
3808 |
|
- Gene Symbol
- PNRC1
- Entrez Gene ID
- 10957
- Gene Name
- proline rich nuclear receptor coactivator 1
- Gene Aliases
- B4-2, PNAS-145, PROL2, PRR2
- Chromosome Location
- Chr. 6: 89080710 - 89085160 on Build GRCh38
- UniGene ID
- Hs.75969
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130722.1 |
- |
1 |
69 |
|
- Gene Symbol
- RCSD1
- Entrez Gene ID
- 92241
- Gene Name
- RCSD domain containing 1
- Gene Aliases
- CAPZIP, MK2S4
- Chromosome Location
- Chr. 1: 167630193 - 167708696 on Build GRCh38
- UniGene ID
- Hs.493867
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_136519.1 |
|
2 |
269 |
|
- Gene Symbol
- RFTN2
- Entrez Gene ID
- 130132
- Gene Name
- raftlin family member 2
- Gene Aliases
- C2orf11, Raftlin-2
- Chromosome Location
- Chr. 2: 197568224 - 197676050 on Build GRCh38
- UniGene ID
- Hs.591615
- Species
- Homo sapiens
- Gene Symbol
- RNF24
- Entrez Gene ID
- 11237
- Gene Name
- ring finger protein 24
- Gene Aliases
- G1L
- Chromosome Location
- Chr. 20: 3927309 - 4015591 on Build GRCh38
- UniGene ID
- Hs.547576
- Species
- Homo sapiens
- Gene Symbol
- SCAND2P
- Entrez Gene ID
- 54581
- Gene Name
- SCAN domain containing 2 pseudogene
- Gene Aliases
- SCAND2
- Chromosome Location
- Chr. 15: 84631460 - 84642463 on Build GRCh38
- UniGene ID
- Hs.658114
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003654.2 |
|
4 |
3054 |
|
NR_004859.1 |
|
5 |
3612 |
|
GenBank mRNA |
BC063836.1 |
- |
4 |
3010 |
|
- Gene Symbol
- SLC35E3
- Entrez Gene ID
- 55508
- Gene Name
- solute carrier family 35 member E3
- Gene Aliases
- BLOV1
- Chromosome Location
- Chr. 12: 68746148 - 68768677 on Build GRCh38
- UniGene ID
- Hs.506011
- Species
- Homo sapiens
- Gene Symbol
- SNX22
- Entrez Gene ID
- 79856
- Gene Name
- sorting nexin 22
- Gene Aliases
- Chromosome Location
- Chr. 15: 64151698 - 64157481 on Build GRCh38
- UniGene ID
- Hs.744250
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK024014.1 |
- |
7 |
1152 |
|
- Gene Symbol
- SPATA5
- Entrez Gene ID
- 166378
- Gene Name
- spermatogenesis associated 5
- Gene Aliases
- AFG2, EHLMRS, SPAF
- Chromosome Location
- Chr. 4: 122923070 - 123319450 on Build GRCh38
- UniGene ID
- Hs.709648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145207.2 |
NP_660208.2
|
16 |
4521 |
|
- Gene Symbol
- TCEA1
- Entrez Gene ID
- 6917
- Gene Name
- transcription elongation factor A1
- Gene Aliases
- GTF2S, SII, TCEA, TF2S, TFIIS
- Chromosome Location
- Chr. 8: 53966554 - 54022456 on Build GRCh38
- UniGene ID
- Hs.491745
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094425.1 |
- |
|
705 |
|
- Gene Symbol
- TLCD2
- Entrez Gene ID
- 727910
- Gene Name
- TLC domain containing 2
- Gene Aliases
- Chromosome Location
- Chr. 17: 1702790 - 1710368 on Build GRCh38
- UniGene ID
- Hs.531005
- Species
- Homo sapiens
- Gene Symbol
- TRPV1
- Entrez Gene ID
- 7442
- Gene Name
- transient receptor potential cation channel subfamily V member 1
- Gene Aliases
- VR1
- Chromosome Location
- Chr. 17: 3565446 - 3609411 on Build GRCh38
- UniGene ID
- Hs.579217
- Species
- Homo sapiens
- Gene Symbol
- TSSK3
- Entrez Gene ID
- 81629
- Gene Name
- testis specific serine kinase 3
- Gene Aliases
- SPOGA3, STK22C, STK22D, TSK3
- Chromosome Location
- Chr. 1: 32362261 - 32364325 on Build GRCh38
- UniGene ID
- Hs.512763
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY048701.1 |
- |
1 |
1887 |
|
- Gene Symbol
- WDR73
- Entrez Gene ID
- 84942
- Gene Name
- WD repeat domain 73
- Gene Aliases
- GAMOS, HSPC264
- Chromosome Location
- Chr. 15: 84640682 - 84654343 on Build GRCh38
- UniGene ID
- Hs.745027
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BM875282.1 |
- |
1 |
86 |
|
- Gene Symbol
- WIPI1
- Entrez Gene ID
- 55062
- Gene Name
- WD repeat domain, phosphoinositide interacting 1
- Gene Aliases
- ATG18, ATG18A, WIPI49
- Chromosome Location
- Chr. 17: 68421281 - 68457524 on Build GRCh38
- UniGene ID
- Hs.463964
- Species
- Homo sapiens
- Gene Symbol
- ZDHHC3
- Entrez Gene ID
- 51304
- Gene Name
- zinc finger DHHC-type containing 3
- Gene Aliases
- DHHC-3, GODZ, ZNF373
- Chromosome Location
- Chr. 3: 44915261 - 44976190 on Build GRCh38
- UniGene ID
- Hs.61430
- Species
- Homo sapiens
- Gene Symbol
- ZNF346
- Entrez Gene ID
- 23567
- Gene Name
- zinc finger protein 346
- Gene Aliases
- JAZ, Zfp346
- Chromosome Location
- Chr. 5: 177022680 - 177081189 on Build GRCh38
- UniGene ID
- Hs.484259
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DB340780.1 |
- |
1 |
417 |
|
- Gene Symbol
- ZNF385D
- Entrez Gene ID
- 79750
- Gene Name
- zinc finger protein 385D
- Gene Aliases
- ZNF659
- Chromosome Location
- Chr. 3: 21418419 - 22372641 on Build GRCh38
- UniGene ID
- Hs.21026
- Species
- Homo sapiens
View Full Product Details