Gene Transcripts

Gene Symbol
B3GAT1
Entrez Gene ID
27087
Gene Name
beta-1,3-glucuronyltransferase 1
Gene Aliases
CD57, GLCATP, GLCUATP, HNK1, LEU7, NK-1, NK1
Chromosome Location
Chr. 11: 134378504 - 134411986 on Build GRCh38
UniGene ID
Hs.381050
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018644.3 NP_061114.2 5 1126
NM_054025.2 NP_473366.1 4 1258
XM_005271506.3 XP_005271563.1 5 1177
XM_011542751.2 XP_011541053.1 6 1389
XM_011542753.2 XP_011541055.1 6 1392
XM_017017550.1 XP_016873039.1 4 1014
XM_017017551.1 XP_016873040.1 5 2951
XM_017017552.1 XP_016873041.1 3 1713
GenBank mRNA AB029396.1 - 5 1040
AK299637.1 - 4 944
AL834455.1 - 2 434
BC010466.1 - 4 1258 4157589
CR457098.1 - 3 865

View Full Product Details