Gene Transcripts

Gene Symbol
Cacna1s
Entrez Gene ID
12292
Gene Name
calcium channel, voltage-dependent, L type, alpha 1S subunit
Gene Aliases
AW493108, Cav1.1, Cchl1a3, fmd, mdg, sj
Chromosome Location
Chr. 1: 136052805 - 136119822 on Build GRCm38
UniGene ID
Mm.4418
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081023.1 NP_001074492.1 5 666
NM_014193.2 NP_055008.2 5 666
GenBank mRNA AK143541.1 - 5 604
L06234.1 - 5 782

View Full Product Details