Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019877.2 | NP_063930.1 | 3 | 228 | |
GenBank mRNA | AB040137.1 | - | 3 | 231 | |
AB047850.1 | - | 3 | 241 | ||
AF237687.1 | - | 3 | 231 | ||
AK003622.1 | - | 3 | 151 | ||
AK187566.1 | - | 2 | 110 | ||
BC025122.1 | - | 3 | 217 | 4187541 |