Gene Transcripts

Gene Symbol
KRTAP6-2
Entrez Gene ID
337967
Gene Name
keratin associated protein 6-2
Gene Aliases
KAP6.2
Chromosome Location
Chr. 21: 30598686 - 30598874 on Build GRCh38
UniGene ID
Hs.553690
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181604.1 NP_853635.1 1 173
GenBank mRNA AB096952.1 - 1 199
BC093915.1 - 1 201
BC093917.1 - 1 201

View Full Product Details