Gene Transcripts

Gene Symbol
KRTAP19-3
Entrez Gene ID
337970
Gene Name
keratin associated protein 19-3
Gene Aliases
GTRHP, KAP19.3
Chromosome Location
Chr. 21: 30491464 - 30491957 on Build GRCh38
UniGene ID
Hs.60480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181609.3 NP_853640.1 1 202
GenBank mRNA AB096944.1 - 1 230
AF139541.1 - 1 202
BC126280.1 - 1 246
BC126282.1 - 1 246

View Full Product Details