Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75732507-75732738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADM
Entrez Gene ID
34
Gene Name
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases
ACAD1, MCAD, MCADH
UniGene
Hs.445040
Gene Chromosome Location
Chr.1: 75724347-75763679 on Build GRCh38
Transcripts
NM_000016.5 NM_001127328.2 NM_001286042.1 NM_001286043.1 NM_001286044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6248415 c.119-1G>T p.? Chr.1: 75732643-75732643 on Build GRCh38
5742726 c.163T>A p.F55I Chr.1: 75732688-75732688 on Build GRCh38
6452793 c.208G>A p.E70K Chr.1: 75732721-75732721 on Build GRCh38
4805641 c.174G>A p.E58E Chr.1: 75732699-75732699 on Build GRCh38
3492381 c.157C>T p.R53C Chr.1: 75732682-75732682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768475053 Chr.1: 75732593-75732593 on Build GRCh38
rs778614270 Chr.1: 75732601-75732601 on Build GRCh38
rs755318101 Chr.1: 75732632-75732632 on Build GRCh38
rs771389892 Chr.1: 75732638-75732638 on Build GRCh38
rs747832368 Chr.1: 75732613-75732613 on Build GRCh38
rs764413858 Chr.1: 75732653-75732653 on Build GRCh38
rs778757873 Chr.1: 75732711-75732711 on Build GRCh38
rs757434857 Chr.1: 75732659-75732659 on Build GRCh38
rs11549026 Chr.1: 75732709-75732709 on Build GRCh38
rs775826164 Chr.1: 75732609-75732609 on Build GRCh38
rs398123072 Chr.1: 75732682-75732682 on Build GRCh38
rs370303161 Chr.1: 75732645-75732645 on Build GRCh38
rs989908970 Chr.1: 75732661-75732661 on Build GRCh38
rs888719915 Chr.1: 75732580-75732580 on Build GRCh38
rs754938068 Chr.1: 75732683-75732683 on Build GRCh38
rs913536924 Chr.1: 75732578-75732578 on Build GRCh38
rs765625793 Chr.1: 75732651-75732651 on Build GRCh38
rs1057517179 Chr.1: 75732698-75732699 on Build GRCh38
rs751647383 Chr.1: 75732656-75732656 on Build GRCh38
rs998228537 Chr.1: 75732637-75732637 on Build GRCh38
rs866259718 Chr.1: 75732692-75732692 on Build GRCh38
rs781177517 Chr.1: 75732663-75732663 on Build GRCh38
rs74090724 Chr.1: 75732624-75732624 on Build GRCh38
rs894821803 Chr.1: 75732540-75732540 on Build GRCh38
rs190806009 Chr.1: 75732553-75732553 on Build GRCh38
rs111347361 Chr.1: 75732660-75732660 on Build GRCh38
rs372633601 Chr.1: 75732623-75732623 on Build GRCh38
rs147559466 Chr.1: 75732652-75732652 on Build GRCh38
rs149308824 Chr.1: 75732712-75732712 on Build GRCh38
rs75795214 Chr.1: 75732561-75732561 on Build GRCh38
rs759997176 Chr.1: 75732650-75732650 on Build GRCh38
rs760185676 Chr.1: 75732628-75732628 on Build GRCh38
rs761432730 Chr.1: 75732632-75732633 on Build GRCh38

View Full Product Details