Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75732678-75732905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADM
Entrez Gene ID
34
Gene Name
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases
ACAD1, MCAD, MCADH
UniGene
Hs.445040
Gene Chromosome Location
Chr.1: 75724347-75763679 on Build GRCh38
Transcripts
NM_000016.5 NM_001127328.2 NM_001286042.1 NM_001286043.1 NM_001286044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5742726 c.163T>A p.F55I Chr.1: 75732688-75732688 on Build GRCh38
6452793 c.208G>A p.E70K Chr.1: 75732721-75732721 on Build GRCh38
5128301 c.229-2A>G p.? Chr.1: 75732851-75732851 on Build GRCh38
4805641 c.174G>A p.E58E Chr.1: 75732699-75732699 on Build GRCh38
5673044 c.245G>T p.W82L Chr.1: 75732881-75732881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374358376 Chr.1: 75732739-75732739 on Build GRCh38
rs752451629 Chr.1: 75732792-75732792 on Build GRCh38
rs770408876 Chr.1: 75732730-75732730 on Build GRCh38
rs571581685 Chr.1: 75732773-75732773 on Build GRCh38
rs1057516480 Chr.1: 75732860-75732860 on Build GRCh38
rs756915078 Chr.1: 75732810-75732810 on Build GRCh38
rs778757873 Chr.1: 75732711-75732711 on Build GRCh38
rs780692978 Chr.1: 75732814-75732814 on Build GRCh38
rs398123074 Chr.1: 75732869-75732869 on Build GRCh38
rs11549026 Chr.1: 75732709-75732709 on Build GRCh38
rs746655981 Chr.1: 75732808-75732808 on Build GRCh38
rs796051897 Chr.1: 75732725-75732725 on Build GRCh38
rs764360637 Chr.1: 75732760-75732760 on Build GRCh38
rs750526774 Chr.1: 75732781-75732781 on Build GRCh38
rs762165843 Chr.1: 75732779-75732779 on Build GRCh38
rs771649282 Chr.1: 75732720-75732720 on Build GRCh38
rs786204566 Chr.1: 75732880-75732881 on Build GRCh38
rs767630461 Chr.1: 75732780-75732780 on Build GRCh38
rs749859620 Chr.1: 75732865-75732865 on Build GRCh38
rs745371854 Chr.1: 75732840-75732840 on Build GRCh38
rs758213925 Chr.1: 75732795-75732795 on Build GRCh38
rs776162089 Chr.1: 75732863-75732863 on Build GRCh38
rs528788578 Chr.1: 75732746-75732746 on Build GRCh38
rs930392145 Chr.1: 75732839-75732839 on Build GRCh38
rs777436128 Chr.1: 75732723-75732723 on Build GRCh38
rs398123073 Chr.1: 75732743-75732743 on Build GRCh38
rs778055470 Chr.1: 75732788-75732788 on Build GRCh38
rs763998240 Chr.1: 75732802-75732805 on Build GRCh38
rs770293389 Chr.1: 75732861-75732861 on Build GRCh38
rs121434280 Chr.1: 75732724-75732724 on Build GRCh38
rs754833969 Chr.1: 75732726-75732726 on Build GRCh38
rs758476299 Chr.1: 75732874-75732874 on Build GRCh38
rs778722578 Chr.1: 75732791-75732791 on Build GRCh38
rs764637486 Chr.1: 75732781-75732781 on Build GRCh38
rs41291506 Chr.1: 75732787-75732787 on Build GRCh38
rs151126827 Chr.1: 75732867-75732867 on Build GRCh38
rs927784210 Chr.1: 75732731-75732731 on Build GRCh38
rs2275378 Chr.1: 75732751-75732751 on Build GRCh38
rs149308824 Chr.1: 75732712-75732712 on Build GRCh38
rs1057516801 Chr.1: 75732742-75732742 on Build GRCh38
rs886042053 Chr.1: 75732866-75732866 on Build GRCh38
rs778891510 Chr.1: 75732735-75732735 on Build GRCh38

View Full Product Details