Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75749314-75749539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADM
Entrez Gene ID
34
Gene Name
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases
ACAD1, MCAD, MCADH
UniGene
Hs.445040
Gene Chromosome Location
Chr.1: 75724347-75763679 on Build GRCh38
Transcripts
NM_000016.5 NM_001127328.2 NM_001286042.1 NM_001286043.1 NM_001286044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5093527 c.810C>T p.D270D Chr.1: 75749508-75749508 on Build GRCh38
4658709 c.769G>A p.E257K Chr.1: 75749467-75749467 on Build GRCh38
7154560 c.795T>A p.G265G Chr.1: 75749505-75749505 on Build GRCh38
6591689 c.797A>G p.D266G Chr.1: 75749507-75749507 on Build GRCh38
1344327 c.798C>T p.D266D Chr.1: 75749508-75749508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762143899 Chr.1: 75749481-75749481 on Build GRCh38
rs752749506 Chr.1: 75749380-75749381 on Build GRCh38
rs749692458 Chr.1: 75749466-75749466 on Build GRCh38
rs377138238 Chr.1: 75749506-75749506 on Build GRCh38
rs1057518408 Chr.1: 75749449-75749449 on Build GRCh38
rs897430591 Chr.1: 75749461-75749461 on Build GRCh38
rs121434274 Chr.1: 75749509-75749509 on Build GRCh38
rs935208265 Chr.1: 75749351-75749351 on Build GRCh38
rs1057516644 Chr.1: 75749418-75749418 on Build GRCh38
rs1052855363 Chr.1: 75749462-75749462 on Build GRCh38
rs947867051 Chr.1: 75749398-75749398 on Build GRCh38
rs767205556 Chr.1: 75749381-75749381 on Build GRCh38
rs752490486 Chr.1: 75749389-75749389 on Build GRCh38
rs201375579 Chr.1: 75749507-75749507 on Build GRCh38
rs121434281 Chr.1: 75749444-75749444 on Build GRCh38
rs552308716 Chr.1: 75749343-75749343 on Build GRCh38
rs774404344 Chr.1: 75749479-75749479 on Build GRCh38
rs373852490 Chr.1: 75749438-75749438 on Build GRCh38
rs878872125 Chr.1: 75749355-75749355 on Build GRCh38
rs763956330 Chr.1: 75749397-75749397 on Build GRCh38
rs746483754 Chr.1: 75749406-75749406 on Build GRCh38
rs1022879056 Chr.1: 75749483-75749483 on Build GRCh38
rs111703511 Chr.1: 75749374-75749374 on Build GRCh38
rs765170454 Chr.1: 75749508-75749508 on Build GRCh38
rs753696849 Chr.1: 75749381-75749381 on Build GRCh38
rs754964318 Chr.1: 75749382-75749382 on Build GRCh38
rs773407237 Chr.1: 75749503-75749503 on Build GRCh38
rs780817518 Chr.1: 75749433-75749433 on Build GRCh38
rs875989867 Chr.1: 75749452-75749452 on Build GRCh38
rs121434276 Chr.1: 75749440-75749440 on Build GRCh38
rs777309510 Chr.1: 75749401-75749401 on Build GRCh38
rs756702790 Chr.1: 75749428-75749428 on Build GRCh38
rs768884003 Chr.1: 75749467-75749467 on Build GRCh38
rs778192900 Chr.1: 75749414-75749414 on Build GRCh38
rs780510026 Chr.1: 75749465-75749465 on Build GRCh38
rs1035989817 Chr.1: 75749453-75749453 on Build GRCh38
rs941714381 Chr.1: 75749418-75749418 on Build GRCh38
rs570815181 Chr.1: 75749485-75749485 on Build GRCh38
rs181598253 Chr.1: 75749387-75749387 on Build GRCh38

View Full Product Details