Gene Transcripts

Gene Symbol
Ext2
Entrez Gene ID
14043
Gene Name
exostoses (multiple) 2
Gene Aliases
AI893565
Chromosome Location
Chr. 2: 93695631 - 93822588 on Build GRCm38
UniGene ID
Mm.4336
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010163.3 NP_034293.2 6 1242
GenBank mRNA AK082572.1 - 6 1224
AK134438.1 - 2 183
AK148090.1 - 6 1232
AK148369.1 - 5 1003
AK150155.1 - 6 1234
AK152265.1 - 1342
AK152840.1 - 1344
AK164342.1 - 6 1228
AK187641.1 - 1 45
AK194704.1 - 1 92
BC006597.1 - 6 1215 3965079
U67837.1 - 6 1217
U72141.1 - 6 1221

View Full Product Details