Gene Transcripts

Gene Symbol
Olr1462
Entrez Gene ID
405993
Gene Name
olfactory receptor 1462
Gene Aliases
Chromosome Location
Chr. 10: 45922017 - 45924132 on Build Rnor_6.0
UniGene ID
Rn.142116
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001097.1 NP_001001097.1 1 252
XM_006246511.2 XP_006246573.1 2 356

View Full Product Details